Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BACE-1 Antibody (SR2142), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | BACE-1 |
---|---|
Clone | SR2142 |
Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1:40 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
BACE-1 Monoclonal antibody specifically detects BACE-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
BACE-1 | |
Western Blot 1:500-1:2000, Immunoprecipitation 1:40 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
APP beta-secretase, ASP2, Aspartyl protease 2, BACEAsp 2, beta-secretase 1, beta-secretase 1 precursor variant 1, beta-site amyloid beta A4 precursor protein-cleaving enzyme, Beta-site amyloid precursor protein cleaving enzyme 1, Beta-site APP cleaving enzyme 1, beta-site APP-cleaving enzyme, beta-site APP-cleaving enzyme 1, EC 3.4.23, EC 3.4.23.46, FLJ90568, HSPC104, KIAA1149, Memapsin-2, Membrane-associated aspartic protease 2, transmembrane aspartic proteinase Asp2 | |
A synthesized peptide derived from human BACE-1 (Uniprot #: P56817) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR2142 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Alzheimers Research, Neurodegeneration, Neuroscience | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
23621 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title