Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BACE-1 Rabbit anti-Human, Mouse, Rat, Clone: 7C5T7, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | BACE-1 |
---|---|
Clone | 7C5T7 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
BACE-1 Monoclonal antibody specifically detects BACE-1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
BACE-1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Alzheimers Research, Neurodegeneration, Neuroscience | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
23621 | |
IgG | |
Affinity purified |
7C5T7 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
APP beta-secretase, ASP2, Aspartyl protease 2, BACEAsp 2, beta-secretase 1, beta-secretase 1 precursor variant 1, beta-site amyloid beta A4 precursor protein-cleaving enzyme, Beta-site amyloid precursor protein cleaving enzyme 1, Beta-site APP cleaving enzyme 1, beta-site APP-cleaving enzyme, beta-site APP-cleaving enzyme 1, EC 3.4.23, EC 3.4.23.46, FLJ90568, HSPC104, KIAA1149, Memapsin-2, Membrane-associated aspartic protease 2, transmembrane aspartic proteinase Asp2 | |
A synthetic peptide corresponding to a sequence within amino acids 402-501 of human BACE-1 (P56817). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title