Learn More
Invitrogen™ BACE1 Biosimilar Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA559131

Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term Genentech anti-BACE1.
BACE1 (beta-secretase) is a key enzyme involved in the production of amyloid beta-peptides (Abeta) found in extracellular amyloid plaques of Alzheimer's disease (AD). In some cases early onset familial AD can be attributed to a Swedish mutation in the amyloid precursor protein (APP), which dramatically enhances the cleavage of this protein by BACE1. This and other genetic and pathological evidence has led to therapeutic approaches that have focused on the inhibition of BACE1 and other APP-cleaving enzymes, such as gamma-secretase stuff. APP is first cleaved by b-secretase, producing a soluble derivative of the protein and a membrane anchored 99-amino acid carboxy-terminal fragment (C99). The C99 fragment serves as substrate for g-secretase to generate the 4 kDa amyloid-b peptide, which is deposited in the brains of all sufferers of Alzheimer's disease. The long-sought b-secretase was recently identified by several groups independently and designated beta-site APP cleaving enzyme (BACE) and aspartyl protease 2 (Asp2). bACE/Asp2 is a novel transmembrane aspartic protease and colocalizes with APP.
Specifications
BACE1 Biosimilar | |
Recombinant Monoclonal | |
Unconjugated | |
Human | |
100 μg | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
1 mg/mL | |
25mM Histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
Protein A | |
RUO | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.