Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BACE1 Rabbit anti-Human, Mouse, Zebrafish, Polyclonal, Proteintech
SDP

Catalog No. 501728266 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8266 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8266 Supplier Proteintech Group Inc Supplier No. 128071AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Accumulation of the amyloid-b (Ab) plaque in the cerebral cortex is a critical event in the pathogenesis Alzheimer′s disease. Ab peptide is generated by proteolytic cleavage of the b-amyloid protein precursor(APP) at b- and g-sites by two proteases. APP is first cleaved by b-secretase, producing a soluble derivative of the protein and a membrane anchored 99-amino acid carboxy-terminal fragment (C99). The C99 fragment serves as substrate for g-secretase to generate the 4 kDa amyloid-b peptide, which is deposited in the brains of all sufferers of Alzheimer′s disease. The long-sought b-secretase was recently identified by several groups independently and designated beta-site APP cleaving enzyme (BACE) and aspartyl protease 2 (Asp2). bACE/Asp2 is a novel transmembrane aspartic protease and colocalizes with APP.

Specifications

Antigen BACE1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.19 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene BACE1
Gene Accession No. P56817, P56818
Gene Alias Asp 2, ASP2, Aspartyl protease 2, BACE, BACE1, Beta secretase 1, HSPC104, KIAA1149, Memapsin 2
Gene Symbols bace1
Host Species Rabbit
Immunogen BACE1 Fusion Protein Ag3559
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23621, 23821, 403005
Target Species Human, Mouse, Zebrafish
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.