Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BACE2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529882
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529882 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529882 Supplier Invitrogen™ Supplier No. PA529882
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Mouse (88%), Rat (88%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

BACE2 or Beta Secretase 2 is responsible for the proteolytic processing of the amyloid precursor protein (APP). BACE2 cleaves APP between residues 690 and 691. This cleavage leads to the generation and extracellular release of beta-cleaved soluble APP. BACE2 is also know to cleave APP between residues 671 and 672. Accumulation of APP is a signature of Alzheimer disease and frequent complication of adult Down syndrome patients.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BACE2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.83 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene BACE2
Gene Accession No. Q6IE75, Q9Y5Z0
Gene Alias 1110059C24Rik; 56 kDa aspartic-like protease; AEPLC; AI850424; ALP56; ARP1; asp 1; ASP1; ASP21; Aspartic-like protease 56 kDa; Aspartyl protease 1; Bace2; BAE2; be; beta secretase 2; Beta-secretase 2; beta-site amyloid beta A4 precursor protein-cleaving enzyme 2; beta-site amyloid precursor protein cleaving enzyme 2; Beta-site APP cleaving enzyme 2; beta-site APP-cleaving enzyme 2; CDA13; CEAP1; Down region aspartic protease; Down syndrome region aspartic protease; DRAP; memapsin 1; memapsin-1; Membrane-associated aspartic protease 1; theta-secretase; transmembrane aspartic proteinase Asp1; UNQ418/PRO852
Gene Symbols BACE2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human BACE2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25825, 288227
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.