Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BACE2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501730871 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-0871 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-0871 Supplier Proteintech Group Inc Supplier No. 163211AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cerebral deposition of amyloid beta peptide is an early and critical feature of Alzheimer′s disease and a frequent complication of Down syndrome. Amyloid beta peptide is generated by proteolytic cleavage of amyloid precursor protein by 2 proteases, one of which is the protein encoded by this gene. This gene localizes to the ′Down critical region′ of chromosome 21. The encoded protein, a member of the peptidase A1 protein family, is a type I integral membrane glycoprotein and aspartic protease. Three transcript variants encoding different isoforms have been described for this gene.

Specifications

Antigen BACE2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene BACE2
Gene Accession No. Q6IE75, Q9JL18, Q9Y5Z0
Gene Alias AEPLC, ALP56, Asp 1, ASP1, ASP21, Aspartic like protease 56 kDa, Aspartyl protease 1, BACE2, BAE2, Beta secretase 2, CDA13, CEAP1, Down region aspartic protease, DRAP, Memapsin 1, Theta secretase, UNQ418/PRO852
Gene Symbols Bace2
Host Species Rabbit
Immunogen BACE2 Fusion Protein Ag9404
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25825, 288227, 56175
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.