Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BACH1 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171814JF549
Description
BACH1 Polyclonal antibody specifically detects BACH1 in Human samples. It is validated for Western BlotSpecifications
BACH1 | |
Polyclonal | |
Western Blot | |
basic region leucine zipper transcriptional regulator BACH1, BTB and CNC homolog 1, BTB and CNC homology 1, basic leucine zipper transcription factor 1, HA2303, transcription regulator protein BACH1, transcription regulator protein10BACH-1 | |
The immunogen for this product maps to a region between residue 350 and 400 of human BTB and CNC Homology 1, Basic Leucine Zipper Transcription Factor 1 using the numbering given in entry NP_001177.1 (GeneID 571). | |
0.1 mL | |
Breast Cancer, Cancer, DNA Repair | |
571 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction