Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BACH1 Antibody (OTI4E11), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270240JF646
Description
BACH1 Monoclonal specifically detects BACH1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
BACH1 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
BACH1 | |
Full length recombinant protein of human BACH1 (NP_996749) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI4E11 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
basic region leucine zipper transcriptional regulator BACH1, BTB and CNC homolog 1, BTB and CNC homology 1, basic leucine zipper transcription factor 1, HA2303, transcription regulator protein BACH1, transcription regulator protein10BACH-1 | |
Mouse | |
Affinity Purified | |
Breast Cancer, Cancer, DNA Repair | |
571.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction