Learn More
Invitrogen™ BACH1 Recombinant Rabbit Monoclonal Antibody (8A8X1)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA551374
Description
Immunogen sequence: TPCQMQLSPA VAKDGSEQIS QKRSECPWLG IRISESPEPG QRTFTTLSSV NCPFISTLST EGCSSNLEIG NDDYVSEPQQ EPCPYACVIS LGDDSETDTE GDSESCSARE QECEVKLPFN AQRIISLSRN DFQSLLKMHK LTPEQLDCIH DIRRRSKNRI AAQRCRKRKL DCIQNLESEI EKLQSEKESL LKERDHILST LGETKQNLTG LCQKVCKEAA LSQEQIQILA KYSAADCPLS FLISEKDKST PDGELALPSI FSLSDRPPAV LPPCARGNSE PGYARGQESQ QMSTATSEQA GPAEQCRQSG GISDFCQQMT DKCTTDE.
BACH1.3 is a transcription factor that belongs to the cap'n'collar type of basic region leucine zipper factor family (CNC-bZip). The encoded protein contains broad complex, tramtrack, bric-a-brac/poxvirus and zinc finger (BTB/POZ) domains, which is atypical of CNC-bZip family members. These BTB/POZ domains facilitate protein-protein interactions and formation of homo and/or hetero oligomers. When this encoded protein forms a heterodimer with MafK, it functions as a repressor of Maf recognition element (MARE) and transcription is repressed. Multiple alternatively spliced transcript variants have been identified for this gene. Some exons of this gene overlap with some exons from the C21orf41 gene, which is transcribed in an opposite orientation to this gene, but which does not seem to encode a protein.
Specifications
BACH1 | |
Recombinant Monoclonal | |
0.07 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
O14867, P97302 | |
BACH1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 410-736 of human BACH1 (NP_001177.1). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
Liquid |
ChIP Assay, ELISA, Western Blot | |
8A8X1 | |
Unconjugated | |
BACH1 | |
6230421P05Rik; AI323795; BACH1; BACH-1; basic region leucine zipper transcriptional regulator BACH1; BTB and CNC homolog 1; BTB and CNC homology 1; BTB and CNC homology 1 transcription factor; BTB and CNC homology 1, basic leucine zipper transcription factor 1; BTB domain and CNC homolog 1; BTBD24; HA2303; Transcription regulator protein BACH1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
12013, 571 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.