Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BACH2 Polyclonal Antibody

BACH2 Polyclonal Antibody
Supplier: Thermo Scientific PA5100792
Description
BACH2 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
| BACH2 | |
| Polyclonal | |
| Unconjugated | |
| BACH2 | |
| Bach2; BTB and CNC homolog 2; BTB and CNC homology 1, basic leucine zipper transcription factor 2; BTB and CNC homology 2; BTB domain and CNC homolog 2; BTBD25; E030004N02Rik; RGD1562865; similar to BTB and CNC homology 1, basic leucine zipper transcription factor 2; transcription regulator protein BACH2; Transcription regulator protein BACH2 (BTB and CNC homolog 2) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12014, 313125, 60468 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P97303, Q9BYV9 | |
| BACH2 | |
| A synthesized peptide derived from human BACH2(Accession Q9BYV9), corresponding to amino acid residues G565-L615. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction