Learn More
BACH2 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305502AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
BACH2 belongs to the bZIP family. It is a transcriptional regulator that acts as repressor or activator. The protein binds to Maf recognition elements (MARE) and play important roles in coordinating transcription activation and repression by MAFK.Specifications
BACH2 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9BYV9 | |
BACH2 | |
Between 791 and 841 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
BACH2 | |
BTB and CNC homolog 2, transcription regulator protein BACH2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
60468 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.