Learn More
Bach2 Polyclonal Antibody, Rockland™

Rabbit Polyclonal Antibody
Supplier: Rockland Immunochemicals 600401H35
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Bach2 antibody is directed against mouse Bach2 (n-terminus) protein. A BLAST analysis was used to suggest 100% homology with Bach2 protein from mouse. Reactivity against homologues from other sources is not known.
BTB and CNC homolog 2 (BACH2) is a transcriptional regulator that acts as repressor or activator. BACH2 binds to Maf recognition elements (MARE). It induces apoptosis in response to oxidative stress through repression of the antiapoptotic factor HMOX1.Specifications
| Bach2 | |
| Polyclonal | |
| Unconjugated | |
| BACH2 | |
| Bach2; BTB and CNC homolog 2; BTB and CNC homology 1, basic leucine zipper transcription factor 2; BTB and CNC homology 2; BTB domain and CNC homolog 2; BTBD25; E030004N02Rik; RGD1562865; similar to BTB and CNC homology 1, basic leucine zipper transcription factor 2; Transcription regulator protein BACH2; Transcription regulator protein BACH2 (BTB and CNC homolog 2) | |
| Rabbit | |
| RUO | |
| 12014 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot | |
| 0.97 mg/mL | |
| 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
| P97303 | |
| BACH2 | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.