Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BACH2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501570596
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-0596 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-157-0596 Supplier Bethyl Laboratories, Inc Supplier No. A305502A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

BACH2 belongs to the bZIP family. It is a transcriptional regulator that acts as repressor or activator. The protein binds to Maf recognition elements (MARE) and play important roles in coordinating transcription activation and repression by MAFK.

Specifications

Antigen BACH2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene BACH2
Gene Accession No. Q9BYV9
Gene Alias BTB and CNC homolog 2, transcription regulator protein BACH2
Gene Symbols BACH2
Host Species Rabbit
Immunogen Between 791 and 841
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60468
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.