Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bacillus anthracis Spore Antigen Monoclonal Antibody (SA26), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA121719
Description
MA1-21719 detects Bacillus anthracis spore antigen in Bacteria samples.
Anthrax toxin is a three-protein exotoxin secreted by virulent strains of the bacterium Bacillus anthracis, the causative agent of anthrax. Anthrax toxin is composed of a cell-binding protein, known as protective antigen (PA), and two enzyme components, called edema factor (EF) and lethal factor (LF). These three protein components act together to impart their physiological effects. Assembled complexes containing the toxin components are endocytosed. In the endosome, the enzymatic components of the toxin translocate into the cytoplasm of a target cell. Once in the cytosol, the enzymatic components of the toxin disrupts various immune cell functions, namely cellular signaling and cell migration. The toxin may even induce cell lysis, as is observed for macrophage cells. Anthrax toxin allows the bacteria to evade the immune system, proliferate, and μgtimately kill the host animal. Research on anthrax toxin also provides insight into the generation of macromolecular assemblies, and on protein translocation, pore formation, endocytosis, and other biochemical processes.
Specifications
| Bacillus anthracis Spore Antigen | |
| Monoclonal | |
| 4.7 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Mouse | |
| Protein G | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG2a |
| ELISA, Western Blot | |
| SA26 | |
| Unconjugated | |
| Anthrax Spore; B. anthracis Spore | |
| Bacillus anthracis spore extract. | |
| 100 μg | |
| Primary | |
| 4° C, do not freeze | |
| Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction