Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bacillus Spore Antigen Polyclonal Antibody, FITC
GREENER_CHOICE

Catalog No. PA173113
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA173113 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA173113 Supplier Invitrogen™ Supplier No. PA173113
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The PA1-73113 antibody reacts with Bacillus species spores. PA1-73113 has been successfully used in Immunofluorescence applications. The PA1-73113 antibody was raised against purified spores of B. cereus (ATCC #11778) + B. subtilis (ATCC #9372).

Bacillus is a genus of rod-shaped, Gram-positive bacteria and a member of the division Firmicutes. Bacillus species are either obligate or facultative aerobes, and test positive for the enzyme catalase. Bacillus includes both free-living (nonparasitic) and parasitic pathogenic species such as B. anthracis, which causes anthrax, and B. cereus, which causes food poisoning. Under stressful environmental conditions, the bacteria can produce oval endospores that allow them to remain in a dormant state for very long periods.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bacillus Spore Antigen
Applications Immunocytochemistry
Classification Polyclonal
Concentration 4-5 mg/mL
Conjugate FITC
Formulation PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2
Gene Alias Anthrax Spore; Bacillus Spore
Host Species Rabbit
Immunogen Purified spores of B. cereus (ATCC #11778) + B. subtilis (ATCC #9372).
Purification Method IgG fraction
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.