Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bacillus subtilis iolG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117572
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117572 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117572 Supplier Thermo Scientific Supplier No. PA5117572
Add to Cart
Add to Cart

Bacillus subtilis iolG Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen Bacillus subtilis iolG
Applications Western Blot
Classification Polyclonal
Concentration 3.695 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene iolG
Gene Accession No. P26935
Gene Symbols iolG
Host Species Rabbit
Immunogen Recombinant Bacillus subtilis Inositol 2-dehydrogenase/D-chiro-inositol 3-dehydrogenase protein (1-344aa).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 937615
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.