Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Bacillus subtilis pbpE Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117578
Description
This antibody has been tested in direct-ELISA.
Probably involved in peptidoglycan modification during cortex synthesis.
Specifications
Bacillus subtilis pbpE | |
Polyclonal | |
Unconjugated | |
pbpE | |
pbpE | |
Recombinant Bacillus subtilis Penicillin-binding protein 4* protein (1-451aa). | |
100 μg | |
Primary | |
Bacteria | |
Antibody | |
IgG |
Western Blot | |
3 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
P32959 | |
Rabbit | |
Protein G | |
RUO | |
938615 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction