Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bacitracin Monoclonal Antibody (4B11)
GREENER_CHOICE

Catalog No. PIMA549733
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549733 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549733 Supplier Invitrogen™ Supplier No. MA549733
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Bacitracin is a mixture of related cyclic polypeptides produced by organisms of the licheniformis group of Bacillus subtilis var Tracy. As a toxic and difficult-to-use antibiotic, bacitracin doesn't work well orally. However, it is very effective topically. Bacitracin is synthesised via the so-called nonribosomal peptide synthetases (NRPSs), which means that ribosomes are not involved in its synthesis. [National Center for Biotechnology Information. PubChem Compound Database; CID=10909430].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bacitracin
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 4B11
Concentration 2.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Alias BACiiM
Host Species Mouse
Immunogen Conjugate of BCT hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.