Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Geobacillus stearothermophilus ATCC™ 7953™
Click to view available options
ATCC®:
7953TM*
Description:
Spore Strip Bacillus stearothermophilus ATCC™ 7953™; 105 spore population
Spore Strip Bacillus stearothermophilus ATCC™ 7953™; 106spore population
Description
Thermo Scientific™ Spore Strips are stabilized microorganism-impregnated strips packaged in individual recloseable plastic bags.
- 10 -6spore population
Specifications
Specifications
| ATCC® | 7953TM* |
| Description | Spore Strip Bacillus stearothermophilus ATCC™ 7953™; 106spore population |
| Format | Spore Strips |
| Product Type | Geobacillus Species |
| Target Organism Class | Gram-positive Rods |
| Test Type | Qualitative QC |
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction