Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bacterial Contamination of Water Test Kit

Click to view available options
:
Bacterial Contamination Of Water
Description
Learn about the sources of bacterial contamination in water, the effects of bacterial contamination of water on human health and ways to minimize the risk of contamination.
- Collect water samples
- Test water samples for possible contamination by coliform and other bacterial strains
- Inoculate agar plates
- Perform a total plate count of bacteria
- Observe bacterial smears under a microscope
Order Info
Class size: 40 students
Specifications
Specifications
| For Use With (Application) | Learn about the sources of bacterial contamination in water, the effects of bacterial contamination of water on human health and ways to minimize the risk of contamination |
| Product Type | Test Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction