Learn More
Bacteriophage M13 Polyclonal Antibody, Rockland™

Rabbit Polyclonal Antibody
$178.35 - $661.74
Specifications
| Antigen | Bacteriophage M13 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | ELISA, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-2356
![]()
|
Rockland Immunochemicals
600401MR5 |
100 μg |
Each for $661.74
|
|
|||||
50-199-2357
![]()
|
Rockland Immunochemicals
600401MR5S |
25 μL |
Each for $178.35
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity purified antibody is directed against Tail virion protein G7P. A BLAST analysis was used to suggest cross-reactivity with the antigen based on 100% homology with the immunizing sequence with Enterobacteria phage M13, Enterobacteria phage f1, and Enterobacteria phage fd.
Tail virion protein G7P (Bacteriophage M13) may initiate with G9P the virion concomitant assembly-budding process, by interacting with the packaging signal of the viral genome. The assembly-budding takes place at the host inner membrane. In turn, G7P and G9P are present at the end of the filamentous virion that emerges first from the bacterial host. Anti-Bacteriophage M13 Antibody is useful for researchers interested in HCP, Host Cell Membranes, Transmembrane Viruses, and E. coli bacteria.Specifications
| Bacteriophage M13 | |
| ELISA, Immunofluorescence, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Virus | |
| Primary | |
| Antibody |
| 1 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
| -20°C, Avoid Freeze/Thaw Cycles |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.