Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bacteriophage M13 Recombinant Mouse Monoclonal Antibody (PS00-01)
GREENER_CHOICE

Catalog No. FEMA550629
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550629 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550629 Supplier Invitrogen™ Supplier No. MA550629
Add to Cart
Add to Cart

Mouse Recombinant Monoclonal Antibody

Positive control: M13 Bacteriophages. Product MA5-50629 was used in an Indirect ELISA.

Tail virion protein G7P (Bacteriophage M13) may initiate with G9P the virion concomitant assembly-budding process, by interacting with the packaging signal of the viral genome. The assembly-budding takes place at the host inner membrane. In turn, G7P and G9P are present at the end of the filamentous virion that emerges first from the bacterial host. Anti-Bacteriophage M13 Antibody is useful for researchers interested in HCP, Host Cell Membranes, Transmembrane Viruses, and E. coli bacteria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bacteriophage M13
Applications ELISA
Classification Recombinant Monoclonal
Clone PS00-01
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide; pH 7.4
Gene Alias Enterobacteria phage M13
Host Species Mouse
Immunogen M13 Bacteriophage.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.