Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Bacteriophage M13 Recombinant Mouse Monoclonal Antibody (PS00-01), HRP
Mouse Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA544933
This item is not returnable.
View return policy
Description
Application notes: Direct ELISA.
Tail virion protein G7P (Bacteriophage M13) may initiate with G9P the virion concomitant assembly-budding process, by interacting with the packaging signal of the viral genome. The assembly-budding takes place at the host inner membrane. In turn, G7P and G9P are present at the end of the filamentous virion that emerges first from the bacterial host. Anti-Bacteriophage M13 Antibody is useful for researchers interested in HCP, Host Cell Membranes, Transmembrane Viruses, and E. coli bacteria.
Specifications
Bacteriophage M13 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.05% BSA, 40% glycerol and no preservative; pH 7.4 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG1 |
ELISA | |
PS00-01 | |
HRP | |
Enterobacteria phage M13 | |
M13 Bacteriophage. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction