Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bacteriophage T7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144557
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144557 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144557 Supplier Invitrogen™ Supplier No. PA5144557
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Bacteriophage T7 Polyclonal Antibody for Western Blot, ELISA

Highly processive DNA-dependent RNA polymerase that catalyzes the transcription of class II and class III viral genes. Recognizes a specific promoter sequence and enters first into an 'abortive phase' where very short transcripts are synthesized and released before proceeding to the processive transcription of long RNA chains. Unwinds the double-stranded DNA to expose the coding strand for templating. Participates in the initiation of viral DNA replication presumably by making primers accessible to the DNA polymerase, thus facilitating the DNA opening. Also plays a role in viral DNA packaging, probably by pausing the transcription at the right end of concatemer junction to allow packaging complex recruitment and beginning of the packaging process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bacteriophage T7
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene 1
Gene Accession No. P00573
Gene Alias T7 RNA polymerase 1
Gene Symbols 1
Host Species Rabbit
Immunogen Recombinant Enterobacteria phage T7 (Bacteriophage T7) 1 protein (710-805aa).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1261050
Target Species Virus
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.