Learn More
Bacteriostatic Water, Sterile, G-Biosciences

Description
- This solution is supplied in borosilicate glass vials for precise dilutions, featuring premium flip cap tops and self-healing butyl rubber stoppers for secure and repeated access.
- It serves as an ideal solvent for dissolving chemicals.
- Manufactured under strict quality control measures, it offers a dependable performance for various laboratory applications.
- This solution is Research Use Only (RUO), not intended for medical use and is not approved for diagnostic or therapeutic applications or in neonates.
Specifications
Specifications
| Percent Purity | >99% |
| Grade | Laboratory |
| Sterility | Sterile |
| Quantity | 3 x 30 mL |
| Chemical Name or Material | Benzyl Alcohol |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.