Learn More
Fisherbrand™ Bacti-Loop™ Tilt Sterilizer
Catalog No. p-7192930 Shop All Fisherbrand Products
Click to view available options
Model:
B1001
B1001-XL
Fisherbrand™ Bacti-Loop™ Tilt Sterilizer has an asbestos-free core element that uses infrared high temperatures 815™C (1500™F).
  • Complete sterilization in seconds without spatter
  • Ideal for platinum loops and needles
  • Tilt model for hands-free sterilization of self-standing vessels
  • Available as XL, with wide chamber for sterilizing larger vessels
  • Completely sterilizes microorganisms within 5 to 7 seconds.
  • Provides instant sterilization of platinum inoculating loops, needles, glass tube/pipette mouths, and various metal and borosilicate glass instruments used in the laboratory.
  • Ceramic heating element is safely enclosed in a perforated
    stainless steel guard chamber, protecting users from accidental
    contact with the extreme temperatures of the core area.
  • Compact, convenient and can be used in anaerobic and aerobic chambers.
Specifications
Model B1001
Product Type Sterilizer
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.