Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BactiSwab™: Wire Shaft Liquid Stuart

Catalog No. p-4525716
Click to view available options
Packaging:
100/Pk.
50/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy

Supplied with a mini-tip swab suitable for ear, nose, throat, eye, male urethral and pediatric specimen collection.

Thermo Scientific™ Remel BactiSwab: Wire Shaft Liquid Stuart is supplied with a mini-tip swab suitable for ear, nose, throat, eye, male urethral and pediatric specimen collection.

Rayon-tipped, plastic-shaft swab(s) are mounted in the cap of a translucent plastic tube with spiral chamber containing a sponge impregnated with liquid transport medium.
  • Straight aluminum shaft
  • Green cap
TRUSTED_SUSTAINABILITY

Specifications

Product Type Tube Kit
Format Tube
Media Type Liquid Stuart
Closure Color Green
Description Liquid Stuarts
For Use With (Application) Healthcare
Packaging 50/Pk.
Shaft Material Wire
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.