Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD™ Bacto™ Dehydrated Agar PROMO

Catalog No. DF014074S24
Click to view available options
:
Bacto Dehydrated Agar; Form: Dehydrated
This item is not returnable. View return policy
This item is not returnable. View return policy

BD Difco™ Bacto™ Dehydrated Agar is a purified solidifying agent used to remove extraneous matter.

  • Extraneous matter, pigmented portions and salts are reduced to minimum
  • Fine, light colored granules
  • Convenient for weighing and handling
  • Determines motility and growth of anaerobes and microaerophiles
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Concentration 1.5%
Additive Agar
Form Dehydrated
For Use With (Application) Determination of Motility and Growth of Anaerobes and Microaerophiles
Packaging Type Container
Quantity 2 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.