Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Bacto™ Soytone
GREENER_CHOICE

Catalog No. p-8843005
Click to view available options
Quantity:
500 g
10 kg

An enzymatic digest of soy that uses an animal origin (AO) (porcine) enzyme for digestion.

  • An enzymatic digest of soy that uses an animal origin (AO) (porcine) enzyme for digestion
  • The naturally high concentrations of vitamins and carbohydrates found in soy make Bacto Soytone a nutritious carbohydrate source for those not concerned about the processing step involving an animal-based material
  • This supplement offers a unique nutritional profile as compared to other soy products
  • It can be a valuable alternative to consider for supplementing both mammalian and microbial processes
  • Store in cool, dry conditions in well-sealed containers at 2°C to 25°C
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance LUO
Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Form Powder
Product Type Dehydrated Culture Media
Format Bottle
For Use With (Application) Enzymatic Hydrolysate of Soybean meal
Product Line Bacto™
Quantity 500 g
Source Animal origin, porcine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.