Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ BactoBeads™ E. coli OP50 (for C.elegans)
SDPSCI_ED

Catalog No. s29021 Shop All EDVOTEK Products
Change view
Click to view available options
Quantity:
5 beads
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S29021 5 beads
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S29021 Supplier Edvotek™ Supplier No. 729
Only null left
Add to Cart
Add to Cart

Non-pathogenic, freeze-dried microbes (bacteria and lower eukaryotes) that are easily cultured for use in the classroom.

  • Includes: 5 Beads
  • Storage: Refrigerator
  • Each bead contains microorganisms, buffer, salts and nutrient broth in an instantly soluble pellet
  • Stable for at least nine months at room temperature, making them easy to ship to school and store in classroom
  • Instantly soluble in water, Luria Broth or any other microbial growth medium
  • Color-coded for easy strain identification
  • Place one BactoBead on the agar plate, watch it dissolve, and streak for isolated colonies

Specifications

Form Freeze-dried
Product Type Escherichia coli
Sterility Non-sterile
For Use With (Application) Classroom
Quantity 5 beads
Species Escherichia coli
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.