Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bad Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $304.50
Specifications
Antigen | Bad |
---|---|
Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry 1:2000, Immunocytochemistry/Immunofluorescence 1:500-1:2000, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:2000 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056080T
![]() |
Novus Biologicals
NB10056080SS |
0.025 mL |
Each for $181.90
|
|
|||||
NB10056080
![]() |
Novus Biologicals
NB10056080 |
0.05 mL |
Each for $304.50
|
|
|||||
Description
Bad Polyclonal specifically detects Bad in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
Bad | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, Tumor Suppressors | |
Whole antisera with 0.02% Sodium Azide | |
572 | |
A full length recombinant protein corresponding to human Bad was used as immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 1:1000-1:2000, Immunohistochemistry 1:2000, Immunocytochemistry/Immunofluorescence 1:500-1:2000, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:2000 | |
Polyclonal | |
Antisera | |
RUO | |
Human | |
BBC2, BBC6, bcl2 antagonist of cell death, BCL2-antagonist of cell death protein, BCL2-associated agonist of cell death, Bcl-2-binding component 6, BCL2-binding component 6, BCL2-binding protein, Bcl2-L-8, BCL2L8bcl2-L-8, Bcl-2-like protein 8, BCL-X/BCL-2 binding protein, Bcl-XL/Bcl-2-associated death promoter | |
BAD | |
IgG | |
Unpurified | |
Bad |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title