Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAD Mouse anti-Human, Mouse, Clone: BYC001, Abnova™

Catalog No. 89933620 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-620 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-620 Supplier Abnova Corporation Supplier No. MAB12571

Mouse monoclonal antibody raised against mouse Bad.

Specifications

Antigen BAD
Applications Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone BYC001
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against mouse Bad.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Immunoprecipitation Western Blot The optimal working dilution should be determined by the end user.
Formulation In 0.1M Tris-glycine, 0.15M NaCl, pH 7.4 (0.05% sodium azide).
Gene Alias AI325008/Bbc2
Gene Symbols Bad
Host Species Mouse
Immunogen Recombinant His6-tag fusion protein corresponding to full length of mouse Bad.
Purification Method Protein G purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12015
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.