Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAD Rabbit anti-Human, Mouse, Rat, PE, Clone: B9, Abnova™

Catalog No. 89335644 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-644 10 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-644 Supplier Abnova Corporation Supplier No. MAB18810
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

BAD Monoclonal antibody specifically detects BAD in Human, Mouse, Rat samples. It is validated for Flow Cyt.

Specifications

Antigen BAD
Applications Flow Cytometry
Classification Monoclonal
Clone B9
Conjugate PE
Description BCL2-associated agonist of cell death
Gene BAD
Gene Alias BBC2, BCL2L8
Host Species Rabbit
Immunogen A synthetic phosphopeptide corresponding to residues surrounding S112 of human BAD
Quantity 10 Reactions
Primary or Secondary Primary
Gene ID (Entrez) 572
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C
Form Liquid
Isotype IgG1 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.