Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAF180/PB1 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10079833G
Description
BAF180/PB1 Polyclonal antibody specifically detects BAF180/PB1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
BAF180/PB1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
BAF180PB1BRG1-associated factor 180, hPB1, MGC156155, MGC156156, polybromo 1, Polybromo-1D, protein polybromo-1 | |
The immunogen recognized by this antibody maps to a region between residue 1639 and 1689 of human polybromo 1 (BRG1-associated factor 180) using the numbering given in entry AAP34197.1 (GeneID 55193). | |
0.1 mL | |
Chromatin Modifiers | |
55193 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction