Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAF53A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | BAF53A |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
BAF53A Polyclonal specifically detects BAF53A in Mouse samples. It is validated for Western Blot.Specifications
BAF53A | |
Polyclonal | |
Rabbit | |
Extracellular Matrix | |
actin-like 6A, actin-like protein 6A, actin-related protein 4,53 kDa BRG1-associated factor A, Actin-related protein Baf53a, Actl6, Arp4, ArpNbeta, BAF complex 53 kDa subunit, Baf53a, BAF53AACTL6, BAF53MGC5382, BRG1-associated factor 53A, hArpN beta, INO80 complex subunit K, INO80KARPN-BETA | |
ACTL6A | |
IgG | |
47 kDa |
Western Blot | |
Unconjugated | |
RUO | |
O96019 | |
86 | |
Synthetic peptides corresponding to ACTL6A (actin-like 6A) The peptide sequence was selected from the N terminal of ACTL6A)(50ug). Peptide sequence VDFPTAIGMVVERDDGSTLMEIDGDKGKQGGPTYYIDTNALRVPRENMEA. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title