Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BAF53A Recombinant Rabbit Monoclonal Antibody (JU40-32)
GREENER_CHOICE

Catalog No. PIMA541247
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541247 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA541247 Supplier Invitrogen™ Supplier No. MA541247
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is predicted to work in Immunoprecipitation at a dilution of 1:10-1:50.

BAF53A is a family member of actin-related proteins (ARPs), which share significant amino acid sequence identity to conventional actins. Both actins and ARPs have an actin fold, which is an ATP-binding cleft, as a common feature. The ARPs are involved in diverse cellular processes, including vesicular transport, spindle orientation, nuclear migration and chromatin remodeling. This gene encodes a 53 kDa subunit protein of the BAF (BRG1/brm-associated factor) complex in mammals, which is functionally related to SWI/SNF complex in S. cerevisiae and Drosophila; the latter is thought to facilitate transcriptional activation of specific genes by antagonizing chromatin-mediated transcriptional repression. Together with beta-actin, it is required for maximal ATPase activity of BRG1, and for the association of the BAF complex with chromatin/matrix. Three transcript variants that encode two different protein isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAF53A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JU40-32
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene Actl6a
Gene Accession No. O96019, Q9Z2N8
Gene Alias 2810432C06Rik; 53 kDa BRG1-associated factor A; actin like 6A; actin-like 6; actin-like 6A; actin-like 6B; actin-like protein 6A; actin-related protein 4; actin-related protein Baf53a; Actl6; Actl6a; ACTL6B; AI851094; ARP4; ArpNbeta; ARPN-BETA; BAF complex 53 kDa subunit; BAF53; Baf53a; BAF53B; BRG1/brm-associated factor 53A; BRG1-associated factor 53A; C79802; hArpN alpha; hArpN beta; INO80 complex subunit K; INO80K; MGC5382; MGC5382 Arp4; RGD1307747
Gene Symbols Actl6a
Host Species Rabbit
Immunogen Recombinant protein within Human ACTL6A aa 185-326.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56456, 86
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.