Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BAF53B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583758
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583758 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583758 Supplier Invitrogen™ Supplier No. PA583758
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CPKADFPTTV GLLAAEEGGG LELEGDKEKK GKIFHIDTNA LHV Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 98%.

Belongs to the chromatin remodeling brain-specific BAF (bBAF) complex, as such plays a role in remodeling mononucleosomes in an ATP-dependent fashion. Belongs to the neuron-specific chromatin remodeling complex (nBAF complex) and is required for postmitotic neural development and dendritic outgrowth. During neural development a switch from a stem/progenitor to a post-mitotic chromatin remodeling mechanism occurs as neurons exit the cell cycle and become committed to their adult state. The transition from proliferating neural stem/progenitor cells to post-mitotic neurons requires a switch in subunit composition of the npBAF and nBAF complexes. As neural progenitors exit mitosis and differentiate into neurons, npBAF complexes which contain ACTL6A/BAF53A and PHF10/BAF45A, are exchanged for homologous alternative ACTL6B/BAF53B and DPF1/BAF45B or DPF3/BAF45C subunits in neuron-specific complexes (nBAF). The npBAF complex is essential for the self-renewal/proliferative capacity of the multipotent neural stem cells. The nBAF complex along with CREST plays a role regulating the activity of genes essential for dendrite growth. ACTL6B/BAF53B is not essential for assembly of the nBAF complex but is required for targeting the complex and CREST to the promoter of genes essential for dendritic growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAF53B
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Actl6b
Gene Accession No. O94805
Gene Alias 53 kDa BRG1-associated factor A; 53 kDa BRG1-associated factor B; actin like 6A; actin like 6B; actin-like 6; actin-like 6A; actin-like 6B; actin-like protein 6A; Actin-like protein 6B; actin-like protein 6B {ECO:0000250; actin-related protein 4; actin-related protein Baf53a; Actin-related protein Baf53b; ACTL6; ACTL6A; Actl6b; actl6b {ECO:0000250; Arp4; ArpNa; ArpNalpha; ArpN-alpha; arpNbeta; ARPN-BETA; BAF complex 53 kDa subunit; BAF53; BAF53A; BAF53B; BRG1-associated factor 53A; BRG1-associated factor 53B; hArpN alpha; hArpN beta; INO80 complex subunit K; INO80K; UniProtKB:O94805}
Gene Symbols Actl6b
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ACTL6B. Recombinant protein control fragment (Product #RP-100177).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51412
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.