Learn More
Invitrogen™ BAF60C Monoclonal Antibody (OTI4G3)

Description
BAF60C Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | BAF60C |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI4G3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SMARCD3 |
| Gene Accession No. | Q6STE5 |
| Gene Alias | 1500001J14Rik; 2210409C08Rik; 60 kDa BRG-1/Brm-associated factor subunit C; BAF60C; BRG1-associated factor 60C; chromatin remodeling complex BAF60C subunit; CRACD3; mammalian chromatin remodeling complex BRG1-associated factor 60C; mBAF60c; MGC111010; Rsc6p; Smarcd3; SWI/SNF complex 60 kDa subunit C; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 3; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 3, isoform 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily D member 3; Swp73-like protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.