Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAFF/BLyS/TNFSF13B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$206.50 - $499.50
Specifications
Antigen | BAFF/BLyS/TNFSF13B |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 0.5-2 ug/ml, Immunohistochemistry-Paraffin reported in scientific literature (PMID 33536561) |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056310T
![]() |
Novus Biologicals
NB10056310SS |
0.025 mg |
Each for $206.50
|
|
|||||
NB10056310
![]() |
Novus Biologicals
NB10056310 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
BAFF/BLyS/TNFSF13B Polyclonal specifically detects BAFF/BLyS/TNFSF13B in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.Specifications
BAFF/BLyS/TNFSF13B | |
Western Blot 0.5-2 ug/ml, Immunohistochemistry-Paraffin reported in scientific literature (PMID 33536561) | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
Q9Y275 | |
10673 | |
This rabbit polyclonal antibody was developed against a C-terminal peptide corresponding to amino acids 254-269 of human TNFSF13B. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis, Cell Cycle and Replication, Cytokine Research, Immunology | |
PBS containing 0.05% BSA with 0.05% Sodium Azide | |
ApoL related ligand TALL-1, B lymphocyte stimulator, BAFFB-cell activating factor, B-cell-activating factor, BLyS, BLYSB-lymphocyte stimulator, CD257, CD257 antigen, Dendritic cell-derived TNF-like molecule, DTL, TALL-1delta BAFF, TALL1Delta4 BAFF, THANK, TNF and ApoL-related leukocyte expressed ligand 1, TNF- and APOL-related leukocyte expressed ligand 1, TNF homolog that activates apoptosis, TNFSF20, tumor necrosis factor (ligand) superfamily, member 13b, tumor necrosis factor (ligand) superfamily, member 20, tumor necrosis factor ligand superfamily member 13B, tumor necrosis factor superfamily, member 13b, tumor necrosis factor-like protein ZTNF4, ZTNF4 | |
TNFSF13B | |
IgG | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title