Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAFFR/TNFRSF13C Rat anti-Mouse, Alexa Fluor 647, Clone: 204406, R&D Systems

Catalog No. FAB1755R100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1755R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1755R100 Supplier R&D Systems Supplier No. FAB1755R100UG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Rat Monoclonal Antibody

BAFFR/TNFRSF13C Monoclonal specifically detects BAFFR/TNFRSF13C in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen BAFFR/TNFRSF13C
Applications Flow Cytometry
Classification Monoclonal
Clone 204406
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q9D8D0
Gene Symbols Tnfrsf13c
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse BAFF R/TNFRSF13C Gly2-Ala71 Accession # Q9D8D0
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115650
Test Specificity Detects mouse BAFF R/TNFRSF13C in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) BAFF R is observed, approximately 10-25% cross-reactivity with recombinant mouse (rm) 4-1BB, rmDR3, rhDR6, rmEDAR, rhHVEM, rmNGF R, rmOX40, and rmTROY is observed, and no cross-reactivity with rmCD27, rmCD30, rmCD40, rmFAS, rmGITR, rmLymphotoxin beta R, rmOPG, rmRANK, rmTNFRI, or rmTNF RII is observed.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.