Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAFF Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984722
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4722 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4722 Supplier Bioss Supplier No. BS2431R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BAFF is a membrane bound cytokine expressed by B cell lineage cells. The soluble form of the cytokine binds to the receptors TNFRSF13B/TACI, TNFRSF17/BCMA, and TNFRSF13C/BAFFR. BAFF is a potent B cell activator, and important for the proliferation and differentiation of B cells. BAFF is also expressed by T lymphocytes and has been shown to promote T cell activation and survival. Isoforms representing alternatively spliced, and glycosylated variants have been identified.

Specifications

Antigen BAFF
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TNFSF13B
Gene Accession No. Q9Y275
Gene Alias ApoL related ligand TALL-1; b cell-activating factor; B lymphocyte stimulator; Baff; B-cell-activating factor; B-lymphocyte stimulator; BLYS; CD257; D8Ertd387e; delta BAFF; Delta4 BAFF; Dendritic cell-derived TNF-like molecule; DTL; RGD1561519; TALL1; TALL-1; THANK; TNF- and APOL-related leukocyte expressed ligand 1; TNF and ApoL-related leukocyte expressed ligand 1; TNF homolog that activates apoptosis; Tnfsf13b; TNFSF20; TNLG7A; tumor necrosis factor (ligand) superfamily member 13b; tumor necrosis factor (ligand) superfamily, member 13b; tumor necrosis factor (ligand) superfamily, member 20; tumor necrosis factor ligand 7A; Tumor necrosis factor ligand superfamily member 13B; Tumor necrosis factor ligand superfamily member 13b, membrane form; Tumor necrosis factor ligand superfamily member 13b, soluble form; tumor necrosis factor superfamily member 13b; tumor necrosis factor-like protein ZTNF4; UNQ401/PRO738; ZTNF4
Gene Symbols TNFSF13B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from mouse BAFF.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10673
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.