Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAFFR/TNFRSF13C Antibody (7J21), Alexa Fluor™ 647, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP222005AF647
Description
BAFFR/TNFRSF13C Monoclonal specifically detects BAFFR/TNFRSF13C in Mouse samples. It is validated for Flow Cytometry, ELISA.Specifications
BAFFR/TNFRSF13C | |
Monoclonal | |
Alexa Fluor 647 | |
B cell-activating factor receptor, BAFF receptor, BAFF-R, BAFFRMGC138235, B-cell-activating factor receptor, BLyS receptor 3, BR3, CD268, CD268 antigen, CVID4, tumor necrosis factor receptor superfamily member 13C, tumor necrosis factor receptor superfamily, member 13C | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, ELISA | |
7J21 | |
Flow Cytometry, ELISA | |
TNFRSF13C | |
Mouse recombinant BAFFR/TNFRSF13C | |
0.1 mL | |
Biologically Active Proteins, Cytokine Research, Immunology, Lipid and Metabolism | |
115650 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction