Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAFFR/TNFRSF13C Antibody (9B9), PerCP, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP280072PCP
This item is not returnable.
View return policy
Description
BAFFR/TNFRSF13C Monoclonal antibody specifically detects BAFFR/TNFRSF13C in Mouse samples. It is validated for Flow CytometrySpecifications
BAFFR/TNFRSF13C | |
Monoclonal | |
PerCP | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry | |
9B9 | |
PBS | |
Y3 cells transfected with mouse BAFFR/TNFRSF13C. | |
0.1 mL | |
Biologically Active Proteins, Cytokine Research, Immunology, Lipid and Metabolism | |
115650 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction