Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAFFR/TNFRSF13C Antibody (BAFFR/1557) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308867
Description
BAFFR/TNFRSF13C Monoclonal specifically detects BAFFR/TNFRSF13C in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
BAFFR/TNFRSF13C | |
Monoclonal | |
1 mg/mL | |
Immunohistochemistry-Paraffin : 1-2 ug/mL | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
BAFFR/1557 | |
Unconjugated | |
10mM PBS | |
Recombinant full-length human BAFFR/TNFRSF13C protein | |
100 μg | |
Biologically Active Proteins, Cytokine Research, Immunology, Lipid and Metabolism | |
115650 | |
Store at -20 to -80C. Avoid freeze-thaw cycles. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction