Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Bag-1 Antibody Blocking Peptide

Click to view available options
Quantity:
0.1 mg
Specifications
Specifications
Gene ID (Entrez) | 573 |
Species | Mouse |
Concentration | 1 mg/mL |
Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB100-56081 |
Gene Alias | BAG family molecular chaperone regulator 1, BAG-1, Bcl-2 associating athanogene-1 protein, BCL2-associated athanogene, Bcl-2-associated athanogene 1, Bcl-2-binding protein, glucocortoid receptor-associated protein RAP46,46-KD, HAP, RAP46 |
Gene Symbol | BAG1 |
Quantity | 0.1 mg |
Research Category | Apoptosis, Cancer, Tumor Suppressors |
Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction