Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bag-1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NB10056081SS
Description
Bag-1 Polyclonal specifically detects Bag-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
Bag-1 | |
Polyclonal | |
Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 | |
BAG family molecular chaperone regulator 1, BAG-1, Bcl-2 associating athanogene-1 protein, BCL2-associated athanogene, Bcl-2-associated athanogene 1, Bcl-2-binding protein, glucocortoid receptor-associated protein RAP46,46-KD, HAP, RAP46 | |
Rabbit | |
Unpurified | |
RUO | |
Primary | |
BAG1 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Whole antisera with 0.02% Sodium Azide | |
BAG1 | |
A synthetic peptide corresponding to amino acids 205-219 (QETERLQSTNLALAE) of mouse BAG-1 was used as immunogen; GenBank no. AAH03722.1. This sequence is 100% conserved in mouse BAG-1L; GenBank no. NP_033866.3. | |
0.025 mL | |
Apoptosis, Cancer, Tumor Suppressors | |
573 | |
Human, Mouse, Rat | |
Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction