Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BAG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520267
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520267 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520267 Supplier Invitrogen™ Supplier No. PA520267
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is PC-3 cell lysate. PA5-20267 can be used with blocking peptide PEP-0382.

The oncogene BCL2 is a membrane protein that blocks a step in a pathway leading to apoptosis or programmed cell death. The protein encoded by this gene binds to BCL2 and is referred to as BCL2-associated athanogene. It enhances the anti-apoptotic effects of BCL2 and represents a link between growth factor receptors and anti-apoptotic mechanisms. At least three protein isoforms are encoded by this mRNA through the use of a non-AUG (CUG) start site, and alternative, downstream, AUG translation initiation sites.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAG1
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Bag1
Gene Accession No. B0K019, Q60739, Q99933
Gene Alias BAG family molecular chaperone regulator 1; Bag1; BAG-1; BCL2 associated athanogene 1; Bcl-2 associating athanogene-1 protein; BCL2-associated athanogene; bcl-2-associated athanogene 1; BCL2-associated athanogene 1; Bcl-2-binding protein; glucocortoid receptor-associated protein RAP46; HAP; Rap46; receptor-associated protein, 46-KD; RP11-326F20.2
Gene Symbols Bag1
Host Species Rabbit
Immunogen A 14 amino acid peptide from near the amino terminus of human BAG-1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12017, 297994, 573
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.