Learn More
Invitrogen™ BAG2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595126
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Testis Tissue, HELA Cell, A549 Cell, A431 Cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
BAG proteins compete with Hip for binding to the Hsc70/Hsp70 ATPase domain and promote substrate release. All the BAG proteins have an approximately 45-amino acid BAG domain near the C terminus but differ markedly in their N-terminal regions. The predicted BAG2 protein contains 211 amino acids. The BAG domains of BAG1, BAG2, and BAG3 interact specifically with the Hsc70 ATPase domain in vitro and in mammalian cells. All 3 proteins bind with high affinity to the ATPase domain of Hsc70 and inhibit its chaperone activity in a Hip-repressible manner.
Specifications
BAG2 | |
Polyclonal | |
Unconjugated | |
BAG2 | |
2610042A13Rik; BAG cochaperone 2; BAG family molecular chaperone regulator 2; BAG2; BAG-2; BAG-family molecular chaperone regulator-2; BC016230; BCL2 associated athanogene 2; Bcl-2-associated athanogene 2; Bcl2-associated athanogene 2; dJ417I1.2; dJ417I1.2 (BAG-family molecular chaperone regulator 2); KIAA0576; MGC149462; RP3-496N17.2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
213539, 690038, 9532 | |
-20°C | |
Lyophilized |
Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
O95816, Q91YN9 | |
BAG2 | |
A synthetic peptide corresponding to a sequence at the N-terminus of human BAG2 (1-14aa MAQAKINAKANEGR). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.