Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BAG2 Recombinant Rabbit Monoclonal Antibody (JE47-57)
GREENER_CHOICE

Catalog No. PIMA536133
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA536133 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA536133 Supplier Invitrogen™ Supplier No. MA536133
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

BAG2 Recombinant Monoclonal Antibody for Flow, Western Blot

BAG proteins compete with Hip for binding to the Hsc70/Hsp70 ATPase domain and promote substrate release. All the BAG proteins have an approximately 45-amino acid BAG domain near the C terminus but differ markedly in their N-terminal regions. The predicted BAG2 protein contains 211 amino acids. The BAG domains of BAG1, BAG2, and BAG3 interact specifically with the Hsc70 ATPase domain in vitro and in mammalian cells. All 3 proteins bind with high affinity to the ATPase domain of Hsc70 and inhibit its chaperone activity in a Hip-repressible manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAG2
Applications Flow Cytometry, Western Blot
Classification Recombinant Monoclonal
Clone JE47-57
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene BAG2
Gene Accession No. O95816
Gene Alias 2610042A13Rik; BAG cochaperone 2; BAG family molecular chaperone regulator 2; BAG2; BAG-2; BAG-family molecular chaperone regulator-2; BC016230; BCL2 associated athanogene 2; Bcl-2-associated athanogene 2; Bcl2-associated athanogene 2; dJ417I1.2; dJ417I1.2 (BAG-family molecular chaperone regulator 2); KIAA0576; MGC149462; RP3-496N17.2
Gene Symbols BAG2
Host Species Rabbit
Immunogen Recombinant protein within Human BAG2 aa 1-118.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9532
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.