Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAG3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP227330SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227330SS 0.025 mL
NBP227330 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227330SS Supplier Novus Biologicals Supplier No. NBP227330SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BAG3 Polyclonal specifically detects BAG3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen BAG3
Applications Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000∼, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000∼, Immunohistochemistry-Frozen 1:1000-1:5000
Formulation 50 ul neat whole antisera with 0.05% Sodium Azide
Gene Accession No. O95817
Gene Alias BAG family molecular chaperone regulator 3, BAG-3, BAG-family molecular chaperone regulator-3, Bcl-2-associated athanogene 3, BCL2-associated athanogene 3, BCL2-binding athanogene 3, Bcl-2-binding protein Bis, BIS, CAIR-1, Docking protein CAIR-1, MGC104307
Gene Symbols BAG3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 276-304 of human BAG-3 was used as immunogen.
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9531
Test Specificity The amino acid sequence used for immunogen is 84% conserved with rat and mouse BAG-3.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.